Learn More
Abcam Human MIP-3 alpha Antibody Pair - BSA and Azide free, 10 x 96 Tests
Catalog No. 502671990
Encompass_Preferred
Human MIP-3 alpha Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human MIP-3 alpha.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1990 Supplier Abcam Supplier No. AB27039610X96TESTS
Human MIP-3 alpha Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human MIP-3 alpha.
The product is subject to the following: Abcam Restricted Use Statement