Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human MIS/AMH ELISA Kit

Catalog No. p-7219525
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH326RBX5 5 x 96 Tests
EH326RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH326RBX5 Supplier Invitrogen™ Supplier No. EH326RBX5
Only null left
Add to Cart
Add to Cart

ELISA

Human MIS/AMH quantitates human MIS/AMH in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human MIS/AMH.

Anti mullerian hormone (AMH) is a member of the TGF beta superfamily. It is secreted as a homodimeric 140 kDa disulfide linked precursor that is cleaved to release the mature 30 kDa homodimer. Originally classified as a fetal testicular hormone that inhibits Mullerian duct development, AMH is expressed post natally by immature Sertoli cells, and to a lesser degree by granulosa cells. AMH plays a role in testicular differentiation and in the regulation of ovarian follicle growth.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Amh; Anti - Mullerian hormone (Mulerian inhibiting substance); Anti-Muellerian hormone; anti-Mullerian hormone; MIF; MIS; Muellerian-inhibiting factor; muellerian-inhibiting substance; Mullerian inhibiting factor; Mullerian inhibiting substance
Assay Range 0.18 to 40 ng/mL
Assay Sensitivity 0.18 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 268
Gene Symbol AMH
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target AMH
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.