Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human MIS RII/AMHR2 ELISA Kit

Catalog No. p-7219524
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH325RBX5 5 x 96 Tests
EH325RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH325RBX5 Supplier Invitrogen™ Supplier No. EH325RBX5
Only null left
Add to Cart
Add to Cart

ELISA

Human MIS RII/AMHR2 quantitates human MIS RII/AMHR2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human MIS RII/AMHR2.

The AMH receptor (AMHR or AMHR2) is a serine/threonine kinase with a single transmembrane domain belonging to the family of type II receptors for TGF-beta-related proteins. Anti-Mullerian hormone (AMH) and its receptor are involved in the regression of Mullerian ducts in male fetuses. Male sex differentiation is mediated by 2 discrete hormones produced by the fetal testis. Testosterone, produced by Leydig cells, virilizes the external genitalia and promotes prostatic growth; anti-Mullerian hormone (AMH) results in regression of Mullerian ducts which would otherwise differentiate into the uterus and fallopian tubes.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AMH type II receptor; AMHR; AMHR2; anti-Muellerian hormone type II receptor; Anti-Muellerian hormone type-2 receptor; anti-Mullerian hormone receptor type 11 SV1; anti-Mullerian hormone receptor type 2; anti-Mullerian hormone receptor type II; anti-Mullerian hormone receptor, type II; anti-Mullerian hormone type 2 receptor; anti-Mullerian hormone type 2 receptor delta 2; anti-Mullerian hormone type 2 receptor delta 9/10; C14; MIS type II receptor; Misiir; MISR2; MISRII; MRII; Muellerian inhibiting substance type II receptor; Mullerian inhibiting substance type II receptor
Assay Range 0.05 to 10 ng/mL
Assay Sensitivity 0.05 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 269
Gene Symbol AMHR2
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target AMHR2
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.