Learn More
Biomatik Corporation Human Mitochondrial Tumor Suppressor 1 (MTUS1) ELISA Kit
Supplier:  Biomatik Corporation EKU12205
Encompass_Preferred
Human Mitochondrial Tumor Suppressor 1 (MTUS1) ELISA Kit for detecting MTUS1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9ULD2. Target Synonyms: ATIP; MP44; MTSG1; ATIP1; ATBP; Microtubule Associated Tumor Suppressor 1; AT2 Receptor-Interacting Protein; Angiotensin-II type 2 receptor-interacting protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8007