Learn More
Biomatik Corporation Human Mitoferrin (MFRN) ELISA Kit
Supplier:  Biomatik Corporation EKU05972
Encompass_Preferred
Human Mitoferrin (MFRN) ELISA Kit for detecting MFRN in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.053ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NYZ2. Gene ID: 51312. Target Synonyms: SLC25A37; MSC; HT015; MSCP; Solute Carrier Family 25 Member 37; Mitochondrial iron transporter 1; Mitochondrial solute carrier protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6035