Learn More
Biomatik Corporation Human Mitogen Activated Protein Kinase 13 (MAPK13) ELISA Kit
Supplier:  Biomatik Corporation EKU05974
Encompass_Preferred
Human Mitogen Activated Protein Kinase 13 (MAPK13) ELISA Kit for detecting MAPK13 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O15264. Gene ID: 5603. Target Synonyms: SAPK4; p38 Delta; Mitogen-activated protein kinase p38 delta; Stress-activated protein kinase 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5945