Learn More
Biomatik Corporation Human Mitogen Activated Protein Kinase Associated Protein 1 (MAPKAP1) ELISA Kit
Supplier:  Biomatik Corporation EKU11284
Encompass_Preferred
Human Mitogen Activated Protein Kinase Associated Protein 1 (MAPKAP1) ELISA Kit for detecting MAPKAP1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.135ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9BPZ7. Gene ID: 79109. Target Synonyms: MIP1; SIN1; SIN1b; SIN1g; Stress-Activated Protein Kinase-Interacting 1; Stress-activated map kinase-interacting protein 1; Target of rapamycin complex 2 subunit MAPKAP1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8174