Learn More
Biomatik Corporation Human Mitogen Activated Protein Kinase 9 (MAPK9) ELISA Kit
Supplier:  Biomatik Corporation EKU11283
Encompass_Preferred
Human Mitogen Activated Protein Kinase 9 (MAPK9) ELISA Kit for detecting MAPK9 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P45984. Gene ID: 5601. Target Synonyms: JNK-55; JNK2; JNK2A; JNK2ALPHA; JNK2B; JNK2BETA; PRKM9; p54a; SAPK; Jun Kinase; Stress-activated protein kinase 1a; c-Jun N-terminal kinase 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8175