Learn More
Biomatik Corporation Human Mitogen Activated Protein Kinase 12 (MAPK12) ELISA Kit
Supplier:  Biomatik Corporation EKU05973
Encompass_Preferred
Human Mitogen Activated Protein Kinase 12 (MAPK12) ELISA Kit for detecting MAPK12 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.072ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P53778. Gene ID: 6300. Target Synonyms: ERK6; PRKM12; p38 Gamma; SAPK3; Stress-activated protein kinase 3; Extracellular signal-regulated kinase 6; Mitogen-activated protein kinase p38 gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8866