Learn More
Biomatik Corporation Human Mitogen Activated Protein Kinase Kinase 2 (MAP2K2) ELISA Kit
Supplier:  Biomatik Corporation EKU12358
Encompass_Preferred
Human Mitogen Activated Protein Kinase Kinase 2 (MAP2K2) ELISA Kit for detecting MAP2K2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P36507. Gene ID: 5605. Target Synonyms: MAPKK2; MEK2; MKK2; PRKMK2; ERK activator kinase 2; Dual specificity mitogen-activated protein kinase kinase 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4959