Learn More
Biomatik Corporation Human Mitotic Arrest Deficient 1 Like Protein 1 (MAD1L1) ELISA Kit
Supplier:  Biomatik Corporation EKU11840
Encompass_Preferred
Human Mitotic Arrest Deficient 1 Like Protein 1 (MAD1L1) ELISA Kit for detecting MAD1L1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y6D9. Gene ID: 8379. Target Synonyms: HsMAD1; MAD1; PIG9; TP53I9; TXBP181; Mitotic spindle assembly checkpoint protein MAD1; Mitotic checkpoint MAD1 protein homolog; Tax-binding protein 181. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5348