Learn More
Sino Biological Anti-Ki67/MKI67 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13180T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Ki67 / MKI67 (rh Ki67 / MKI67; Catalog#13180-H09E; P46013-2; Met1-Pro120). Ki67 / MKI67 specific IgG was purified by Human Ki67 / MKI67 affinity chromatography.
Catalog No. 50-241-1998