Learn More
Sino Biological Anti-Ki67/MKI67 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100130T02100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Ki67/MKI67, and purified by antigen affinity chromatography.
Catalog No. 50-255-5530