Learn More
Sino Biological Anti-CD10/Neprilysin Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 10805MM05P100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD10 / Neprilysin / MME (rh CD10 / Neprilysin / MME; Catalog#10805-H07H; NP_000893.2; Tyr52-Trp750) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-241-1380