Learn More
Sino Biological Human MME MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10805MM13100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD10/Neprilysin/MME (rh CD10/Neprilysin/MME; Catalog#10805-H07H; NP_000893.2; Tyr52-Trp750). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2758