Learn More
Sino Biological Anti-CD10/Neprilysin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10805T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD10/Neprilysin/MME (rh CD10/Neprilysin/MME; Catalog#10805-H07H; NP_000893.2; Tyr52-Trp750). CD10/Neprilysin/MME specific IgG was purified by Human CD10/Neprilysin/MME affinity chromatography.
Catalog No. 50-241-1484