Learn More
Sino Biological Human MME Protein 50ug (Sino Biological)
Supplier:  Sino Biological 10805H01H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Tyr 52-Trp 750) of human membrane metallo-endopeptidase (NP_000893.2) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-237-1714