Learn More
Raybiotech Inc HUMAN MMP-1 ELISA
Supplier:  Raybiotech Inc ELHMMP11
Encompass_Preferred
RayBio Human MMP-1 ELISA Kit for cell culture supernatants, plasma, and serum samples. Pre-coated 96-well microplate kit with additional reagents. Detection Range: 8 pg/ml - 18000 pg/ml
Catalog No. 50-104-4111