Learn More
Abcam Human MMP-12 ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB2138111X96TESTS
Encompass_Preferred
Human MMP-12 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human MMP-12 in Human in Cell culture supernatant, Heparin Plasma, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2204558