Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human MMP-14 ELISA Kit

Catalog No. EEL068
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEL068 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEL068 Supplier Invitrogen™ Supplier No. EEL068
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Human MMP-14 ELISA quantitates MMP-14 in serum, plasma, and other biological fluids The Human MMP-14 (Matrix Metalloproteinase 14) solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the binding of the second (detector) antibody to the target on a different epitope from the capture antibody. An antibody conjugated with enzyme binds the formed sandwich. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. However, the protein encoded by this gene is a member of the membrane-type MMP (MT-MMP) subfamily; each member of this subfamily contains a potential transmembrane domain suggesting that these proteins are expressed at the cell surface rather than secreted. This protein activates MMP2 protein, and this activity may be involved in tumor invasion.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P50281
Assay Range 0.63-40 ng/mL
Assay Sensitivity 0.38 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Matrix metalloproteinase
Sample Type Plasma, Serum
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias AI325305; cb386; EC 3.4.24.80; matrix metallo protease; matrix metallopeptidase 14; matrix metallopeptidase 14 (membrane-inserted); matrix metallopeptidase 14a (membrane-inserted); matrix metalloproteinase 14 (membrane-inserted); matrix metalloproteinase 14 (membrane-inserted) alpha; matrix metalloproteinase 14 membrane-inserted; matrix metalloproteinase 14, membrane-inserted; matrix metalloproteinase 14a (membrane-inserted); matrix metalloproteinase-14; matrix metalloproteinase-14a; membrane type 1 metalloprotease; membrane type 1-matrix metalloproteinase; Membrane type 1-MMP; membrane-type matrix metalloproteinase 1; membrane-type matrix metalloproteinase 1 alpha; membrane-type-1 matrix metalloproteinase; MMP; MMP14; MMP-14; mmp14a; MMPs; MMP-X MT-MMP; MMP-X1; MT1MMP; MT1-MMP; MT1-MMP-alpha; Mtmmp; MT-MMP; MT-MMP 1; MT-MMP MT1-MMP; MTMMP1; MT-MMP-1; MT-MMPalpha; sabe; sb:cb386; type 1 matrix metalloprotease 14; WNCHRS; wu:fb72b09; zMT-MMPalpha; zMT-MMP-alphaAI325305; cb386; EC 3.4.24.80; matrix metallo protease; matrix metallopeptidase 14; matrix metallopeptidase 14 (membrane-inserted); matrix metallopeptidase 14a (membrane-inserted); matrix metalloproteinase 14 (membrane-inserted); matrix metalloproteinase 14 (membrane-inserted) alpha; matrix metalloproteinase 14 membrane-inserted; matrix metalloproteinase 14, membrane-inserted; matrix metalloproteinase 14a (membrane-inserted); matrix metalloproteinase-14; matrix metalloproteinase-14a; membrane type 1 metalloprotease; membrane type 1-matrix metalloproteinase; Membrane type 1-MMP; membrane-type matrix metalloproteinase 1; membrane-type matrix metalloproteinase 1 alpha; membrane-type-1 matrix metalloproteinase; MMP; MMP14; MMP-14; mmp14a; MMPs; MMP-X MT-MMP; MMP-X1; MT1MMP; MT1-MMP; MT1-MMP-alpha; Mtmmp; MT-MMP; MT-MMP 1; MT-MMP MT1-MMP; MTMMP1; MT-MMP-1; MT-MMPalpha; sabe; sb:cb386; type 1 matrix metalloprotease 14; WNCHRS; wu:fb72b09; zMT-MMPalpha; zMT-MMP-alpha;
Gene ID (Entrez) 4323
Gene Symbol MMP14
Interassay CV <10%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 100 μL, Serum 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Refer to product documentation for component specific storage temperature.
Target MMP14
Test Time 1 hr 20 min
Total Assay Time 3 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.