Learn More
ACROBiosystems Human MMP-7, His Tag (MM7-H5249) is expressed from human 293 cells (HEK293). It contains AA Leu 18 - Lys 267 (Accession # P09237-1).
Supplier:  ACROBiosystems MM7H524950UG
Encompass_Preferred
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 29.7 kDa.
Catalog No. 50-223-0938