Learn More
Raybiotech Inc C-Series Human Matrix Metalloproteinase Antibody Array 1 Kit.
Catalog No. 501948620
Encompass_Preferred
Detects 10 Human MMPs and TIMPs. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: MMP-1, MMP-2, MMP-3, MMP-8, MMP-9, MMP-10, MMP-13, TIMP-1, TIMP-2, TIMP-4
Catalog No. 50-194-8620 Supplier Raybiotech Inc Supplier No. AAHMMP12
Detects 10 Human MMPs and TIMPs. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: MMP-1, MMP-2, MMP-3, MMP-8, MMP-9, MMP-10, MMP-13, TIMP-1, TIMP-2, TIMP-4