Learn More
Raybiotech Inc Human MMP Array Q1, 22 Sample Kit
Supplier:  Raybiotech Inc QAHMMP12
Encompass_Preferred
Quantibody® Human Matrix Metalloproteinase Antibody Array 1 Kit. Detects 10 Human MMPs. Suitable for all liquid sample types.
Catalog No. NC3718207