Learn More
Raybiotech Inc Human MMP Array Q1 8 Sample Kit
Catalog No. NC3718201
Encompass_Preferred
Quantibody® Human Matrix Metalloproteinase Antibody Array 1 Kit. Detects 10 Human MMPs. Suitable for all liquid sample types.
Catalog No. NC3718201 Supplier Raybiotech Inc Supplier No. QAHMMP11
Quantibody® Human Matrix Metalloproteinase Antibody Array 1 Kit. Detects 10 Human MMPs. Suitable for all liquid sample types.