Learn More
Sino Biological Anti-MMP-1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10532MM8720
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human MMP1 (rh MMP1; Catalog#10532-H08H; NP_002412.1; Met1-Asn469). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8493