Learn More
Sino Biological Anti-MMP-1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10532T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human MMP1 (rh MMP1; Catalog#10532-H08H; NP_002412.1; Met1-Asn469). MMP1 specific IgG was purified by Human MMP1 affinity chromatography.
Catalog No. 50-255-5270