Learn More
Sino Biological Recombinant Human MMP-1 Protein (His Tag), HPLC-verified 100 µg
Supplier:  Sino Biological 10532H08H100
Encompass_Preferred
A DNA sequence encoding the pro form of human MMP1 enzyme (NP_002412.1) precursor (Met 1-Asn 469) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-224-7198