Learn More
Sino Biological Recombinant Human MMP-10/MMP10 Protein (His Tag)
Supplier:  Sino Biological 10249H07H100
Encompass_Preferred
A DNA sequence encoding the Human MMP10 (P09238) (Phe99-Cys476) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-241-1072