Learn More
Biomatik Corporation Human MMP14 (Matrix metalloproteinase-14) ELISA Kit
Supplier:  Biomatik Corporation EKF58682
Encompass_Preferred
Human MMP14 (Matrix metalloproteinase-14) ELISA Kit for detecting MMP14 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P50281. Target Synonyms: MMP14; MMP-14; MMP-X1; MT1-MMP; MT-MMP1; matrix metallopeptidase 14; membrane-inserted; membrane type 1 metalloprotease; Membrane-type matrix metalloproteinase 1; Membrane-type-1 matrix metalloproteinase; MT1MMP; MTMMP1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9164