Learn More
Abcam Human MMP2 ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1006061X96TESTS
Encompass_Preferred
Human MMP2 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human MMP2 in Human in Plasma, Cell culture supernatant, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2310136