Learn More
Sino Biological Recombinant Anti-MMP-2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10082R029100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human MMP-2 (rh MMP-2; Catalog#10082-HNAH; NP_004521.1; Met 1-Cys660).
Catalog No. 50-240-8430