Learn More
Sino Biological Anti-MMP-2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10082RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human MMP2 (rh MMP-2; Catalog#10082-HNAH; NP_004521.1; Met 1-Cys 660). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8299