Learn More
Sino Biological Human MMP2 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10082T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human MMP-2 / MMP2 (rh MMP-2 / MMP2; Catalog#10082-HNAH; NP_004521.1; Met1-Cys660). MMP-2 / MMP2 specific IgG was purified by Human MMP-2 / MMP2 affinity chromatography.
Catalog No. 50-237-2069