Learn More
BIOHIPPO Human MMP23B Microsample
SDP
Catalog No. 5000314225
Encompass
Human MMP23B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MMP23B in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 29 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MMP23B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MMP23B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MMP23B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14225 Supplier BIOHIPPO Supplier No. BHE15212156
Human MMP23B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MMP23B in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 29 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MMP23B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MMP23B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MMP23B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color