Learn More
Sino Biological Human MMP3 ELISA Pair Set 15PP (Sino Biological)
Catalog No. 502373458
Encompass_Preferred
Each vial contains 174 ng of recombinant Human MMP-3 / MMP3. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 3000 pg/mL is recommended.
Catalog No. 50-237-3458 Supplier Sino Biological Supplier No. SEK1046715
Each vial contains 174 ng of recombinant Human MMP-3 / MMP3. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 3000 pg/mL is recommended.