Learn More
Sino Biological Anti-MMP-3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10467T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human MMP-3 / MMP3 (rh MMP-3 / MMP3; Catalog#10467-HNAE; AAA36321.1; Tyr18-Thr272). MMP-3 / MMP3 specific IgG was purified by Human MMP-3 / MMP3 affinity chromatography.
Catalog No. 50-240-9261