Learn More
BioVendor, LLC QuickZyme Human MMP-7 Activity Assay Kit 96-Assays
SDP
Supplier:  BioVendor, LLC QZBMMP7H
Encompass
QuickZyme Human MMP-7 Activity Assay Kit, 96-Assays. Enables measurement of both active human MMP-2, as well as (pro)MMP-7 (activated on the plate by APMA) in a broad range of sample types (ex. serum, urine, plasma, tissue homogenates, cell culture conditioned medium). This 96-well plate format assay uses a modified pro-enzyme as a substrate, which upon activation is able to release color from a chromogenic peptide substrate. This multiplication step provides a unique assay sensitivity. Cal. range: 0-16 ng/ml. Detection limit: 0.03 ng/ml (3 hr incubation), 0.009 ng/ml (6 hr incub.), 0.004 ng/ml (25 h incub.). Sample vol.: 10-100 ul. All standards, buffers and plates are included: Pre-coated 96-well microplate, assay buffer, APMA, pro-detection enzyme, peptide substrate, wash buffer, human MMP-7 standard. Easy, sensitive, & quantitative replacement for zymography. Synonyms: Matrix metalloproteinase-7, MMP7, Matrilysin, Matrilysin-1, Matrin, Pump-1 protease, Uterine metalloproteinase.
Catalog No. 50-206-9118