Learn More
Sino Biological Anti-MMP-8 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10254T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human MMP-8 (rh MMP-8; Catalog#10254-H08H; NP_002415.1; Met1-Gly467). MMP-8 specific IgG was purified by Human MMP-8 affinity chromatography.
Catalog No. 50-241-1602