Learn More
Sino Biological Recombinant Human MMP-9 Protein (His Tag), HPLC-verified 1 mg
Supplier:  Sino Biological 10327H08H1
Encompass_Preferred
A DNA sequence encoding the pro form of human MMP9 enzyme (NP_004985.2) (Met 1-Asp 707) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-224-6719