Learn More
Sino Biological Human MMP9 ELISA Pair Set 5PP (Sino Biological)
Catalog No. 502371742
Encompass_Preferred
Each vial contains 80 ng of recombinant Human MMP9. Reconstitute with 1 mL Dilution Buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in Dilution Buffer, and a high standard of 8000 pg/mL is recommended.
Catalog No. 50-237-1742 Supplier Sino Biological Supplier No. SEKA103275
Each vial contains 80 ng of recombinant Human MMP9. Reconstitute with 1 mL Dilution Buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in Dilution Buffer, and a high standard of 8000 pg/mL is recommended.