Learn More
Sino Biological Human MMP9 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 10327R04320
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human MMP9 / MMP-9 / CLG4B (rh MMP9 / MMP-9 / CLG4B; Catalog#10327-H08H; NP_004985.2; Met1-Asp707).
Catalog No. 50-237-3091