Learn More
Sino Biological Human MMP9 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10327T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human MMP-9 (rh MMP-9; Catalog#10327-H08H; NP_004985.2; Met1-Asp707). MMP-9 specific IgG was purified by Human MMP-9 affinity chromatography.
Catalog No. 50-237-3682