Learn More
Description
RP-75653 contains purified recombinant human MMP-9 protein with an N-terminal hexahistidine tag. RP-75653 may be used as a positive control in many immunological assays.
Specifications
Specifications
| Accession Number | P14780 |
| Concentration | 0.05 mg/mL |
| For Use With (Application) | Bioactivity, Western Blot |
| Formulation | Laemmli with no preservative; pH 6.8 |
| Gene ID (Entrez) | 4318 |
| Molecular Weight (g/mol) | 42.03 kDa |
| Name | Human MMP-9 |
| pH Range | 6.8 |
| Purification Method | Purified |
| Quantity | 10 μg |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.