Learn More
Raybiotech Inc Human MMR (CD206/MRC1) ELISA Kit
Supplier:  Raybiotech Inc ELHMMR1
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 0.41 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Human Macrophage Mannose Receptor (MRC1 / CD206)
Catalog No. 50-194-9564