Learn More
Techcyte Human Modified Acid Fast O&P Fecal Analysis
Description
- Assist laboratory technologists in the primary screening of Modified Acid-Fast prepared human fecal microscope slides
- Software locates, counts, and pre-classifies parasites, including cryptosporidium sp. oocysts and cyclospora cayetanensis oocysts, and yeasts from a digital image
- Intended for human fecal samples that have been prepared and stained using a modified acid fast staining method
Specifications
Specifications
| Product Type | Detection Software |
| For Use With (Application) | Analyzing Data |
| Operating System | Microsoft Windows 10 IoT 64-bit |
For investigational use only in the US.