Learn More
ACROBiosystems Human MOG 25ug
Catalog No. 502534418
Encompass_Preferred
Biotinylated Human MOG Protein HisAvitag (MOG-H82E5) is expressed from human 293 cells (HEK293). It contains AA Gly 30 - Gly 154 (Accession # Q16653-1).
Catalog No. 50-253-4418 Supplier ACROBiosystems Supplier No. MOGH82E525UG
Biotinylated Human MOG Protein HisAvitag (MOG-H82E5) is expressed from human 293 cells (HEK293). It contains AA Gly 30 - Gly 154 (Accession # Q16653-1).