Learn More
ACROBiosystems Human MOG Protein, His Tag (MALS verified)
Supplier:  ACROBiosystems MOGH52H3100UG
Encompass_Preferred
Human MOG, His Tag (MOG-H52H3) is expressed from human 293 cells (HEK293). It contains AA Gly 30 - Gly 154 (Accession # Q16653-1).
Catalog No. 50-233-1105