Learn More
Kingfisher Biotech Inc Human/Monkey CD40 Ligand (CD40LG) Recombinant Protein (Yeast, Endotoxin - Free) - 100 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0989HY100
Encompass_Preferred
The Human & Monkey CD40 Ligand yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Human & Monkey CD40 Ligand applications are for cell culture, ELISA standard, and Western Blot Control. The Human & Monkey CD40 Ligand yeast-derived recombinant protein can be purchased in multiple sizes. Human & Monkey CD40 Ligand Specifications: (Molecular Weight: 16.2 kDa) (Amino Acid Sequence: MQKGDQNPQI AAHVISEASS KTTSVLQWAE KGYYTMSNNL VTLENGKQLT VKRQGLYYIY AQVTFCSNRE ASSQAPFIAS LCLKSPGRFE RILLRAANTH SSAKPCGQQS IHLGGVFELQ PGASVFVNVT DPSQVSHGTG FTSFGLLKL (149)) (Gene ID: 959). For research use only. Made in the USA
Catalog No. 50-241-3167