Learn More
Biomatik Corporation Human Monoacylglycerol-O-acyltransferase 2 (MOGAT2) ELISA Kit
Supplier:  Biomatik Corporation EKU05981
Encompass_Preferred
Human Monoacylglycerol-O-acyltransferase 2 (MOGAT2) ELISA Kit for detecting MOGAT2 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q3SYC2. Gene ID: 80168. Target Synonyms: MGAT2; DGAT2L5; DC5; 2-Acylglycerol O-Acyltransferase 2; Diacylglycerol O-acyltransferase candidate 5; Diacylglycerol acyltransferase 2-like protein 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5038