Learn More
Biomatik Corporation Human Monoamine Oxidase A (MAOA) ELISA Kit
Supplier:  Biomatik Corporation EKU11288
Encompass_Preferred
Human Monoamine Oxidase A (MAOA) ELISA Kit for detecting MAOA in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.119ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P21397. Gene ID: 4128. Target Synonyms: MAO-A; Amine oxidase [flavin-containing] A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4448